Assay Details
| Gene Symbol: Blvra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026678.4 | NP_080954.4 | 5 - 6 | 543 | 93 | |
| GenBank mRNA | AK002231.1 | - | 4 - 5 | 482 | 93 | |
| AK010847.1 | - | 5 - 6 | 498 | 93 | ||
| AK172620.1 | - | 5 - 6 | 508 | 93 | ||
| BC052146.1 | - | 5 - 6 | 533 | 6588485 | 93 | |
| CA983696.1 | - | 3 - 4 | 278 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Blvra
Genomic Map