Assay Details
| Gene Symbol: Wdr20rt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027614.1 | NP_081890.1 | 1 - 2 | 1976 | 59 | |
| GenBank mRNA | AK015014.1 | - | 1 - 2 | 1976 | 59 | |
| AK015210.1 | - | 1426 | 59 | |||
| BC050778.1 | - | 1 - 2 | 1763 | 6743775 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr20rt
Genomic Map