Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027727.2 | NP_082003.1 | 2 - 3 | 404 | 79 | |
| GenBank mRNA | AK016957.1 | - | 2 - 3 | 333 | 79 | |
| AK049909.1 | - | 2 - 3 | 369 | 79 | ||
| AK080149.1 | - | 2 - 3 | 381 | 79 | ||
| AK081242.1 | - | 2 - 3 | 396 | 79 | ||
| AK149628.1 | - | 2 - 3 | 378 | 79 | ||
| AK149879.1 | - | 2 - 3 | 379 | 79 | ||
| AK149891.1 | - | 2 - 3 | 378 | 79 | ||
| AK154794.1 | - | 2 - 3 | 393 | 79 | ||
| BC038276.1 | - | 2 - 3 | 406 | 4458792 | 79 | |
| BC092283.1 | - | 2 - 3 | 390 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049909.1 transcripts.
Set Membership: |
|

Dym
Genomic Map