Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027727.2 | NP_082003.1 | 3 - 4 | 454 | 71 | |
| GenBank mRNA | AK016957.1 | - | 3 - 4 | 383 | 71 | |
| AK049909.1 | - | 3 - 4 | 419 | 71 | ||
| AK080149.1 | - | 3 - 4 | 431 | 71 | ||
| AK081242.1 | - | 3 - 4 | 446 | 71 | ||
| AK149628.1 | - | 3 - 4 | 428 | 71 | ||
| AK149879.1 | - | 3 - 4 | 429 | 71 | ||
| AK149891.1 | - | 3 - 4 | 428 | 71 | ||
| AK154794.1 | - | 3 - 4 | 443 | 71 | ||
| BC038276.1 | - | 3 - 4 | 456 | 4458792 | 71 | |
| BC092283.1 | - | 3 - 4 | 440 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049909.1 transcripts.
Set Membership: |
|

Dym
Genomic Map