Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027727.2 | NP_082003.1 | 13 - 14 | 1715 | 82 | |
| GenBank mRNA | AK049909.1 | - | 13 - 14 | 1680 | 82 | |
| AK154794.1 | - | 13 - 14 | 1704 | 82 | ||
| AK198427.1 | - | 1 - 2 | 29 | 82 | ||
| AK215509.1 | - | 1 - 2 | 29 | 82 | ||
| BC018220.1 | - | 9 - 10 | 1075 | 3985017 | 82 | |
| BC038276.1 | - | 13 - 14 | 1717 | 4458792 | 82 | |
More Information
Set Membership: |
|

Dym
Genomic Map