Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 3 - 4 | 633 | 148 | |
| GenBank mRNA | AK014813.1 | - | 3 - 4 | 603 | 148 | |
| AK044360.1 | - | 3 - 4 | 634 | 148 | ||
| AK157352.1 | - | 3 - 4 | 602 | 148 | ||
| BC138937.1 | - | 3 - 4 | 542 | 8861959 | 148 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK014813.1,AK044360.1 transcripts.
Set Membership: |
|

Esyt2
Genomic Map