Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 14 - 15 | 1620 | 128 | |
| GenBank mRNA | AK014813.1 | - | 16 - 17 | 1730 | 128 | |
| AK044360.1 | - | 14 - 15 | 1622 | 128 | ||
| AK157352.1 | - | 14 - 15 | 1589 | 128 | ||
| BC040392.1 | - | 4 - 5 | 303 | 128 | ||
| BC052440.1 | - | 4 - 5 | 311 | 128 | ||
| BC054797.1 | - | 1 - 2 | 273 | 128 | ||
| BC059230.1 | - | 4 - 5 | 311 | 128 | ||
| BC138937.1 | - | 14 - 15 | 1529 | 8861959 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Esyt2
Genomic Map