Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 16 - 17 | 1852 | 74 | |
| GenBank mRNA | AK014813.1 | - | 18 - 19 | 1962 | 74 | |
| AK044360.1 | - | 16 - 17 | 1854 | 74 | ||
| AK157352.1 | - | 16 - 17 | 1821 | 74 | ||
| BC040392.1 | - | 6 - 7 | 535 | 74 | ||
| BC052440.1 | - | 6 - 7 | 543 | 74 | ||
| BC054797.1 | - | 3 - 4 | 505 | 74 | ||
| BC059230.1 | - | 6 - 7 | 543 | 74 | ||
| BC138937.1 | - | 16 - 17 | 1761 | 8861959 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Esyt2
Genomic Map