Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 18 - 19 | 2323 | 69 | |
| GenBank mRNA | AK014813.1 | - | 20 - 21 | 2433 | 69 | |
| AK044360.1 | - | 18 - 19 | 2325 | 69 | ||
| AK157352.1 | - | 18 - 19 | 2292 | 69 | ||
| BC040392.1 | - | 8 - 9 | 1006 | 69 | ||
| BC052440.1 | - | 8 - 9 | 1014 | 69 | ||
| BC054797.1 | - | 5 - 6 | 976 | 69 | ||
| BC059230.1 | - | 8 - 9 | 1014 | 69 | ||
| BC138937.1 | - | 18 - 19 | 2232 | 8861959 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Esyt2
Genomic Map