Assay Details
| Gene Symbol: Rbm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029397.3 | NP_083673.3 | 2 - 3 | 263 | 75 | |
| NM_170598.2 | NP_733486.2 | 2 - 3 | 231 | 75 | ||
| GenBank mRNA | AF393215.1 | - | 2 - 3 | 226 | 75 | |
| AF393216.1 | - | 2 - 3 | 194 | 75 | ||
| AK035014.1 | - | 2 - 3 | 221 | 75 | ||
| AK080772.1 | - | 2 - 3 | 221 | 75 | ||
| AK183965.1 | - | 2 - 3 | 100 | 75 | ||
| BC052473.1 | - | 2 - 3 | 206 | 6833669 | 75 | |
| Gene Symbol: Cpne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC058662.1 | - | 2 - 3 | 167 | 75 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF393215.1,AF393216.1,AK035014.1,AK080772.1,BC052473.1,NM_029397.3,NM_170598.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769575 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769581 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076001 | BC052473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076001 | BC026891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076001 | BC027810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076001 | AK085111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076001 | AK166833.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112144 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076001 | BC052473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076001 | BC026891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076001 | BC027810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076001 | AK085111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076001 | AK166833.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276652 | BC052473.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276652 | BC026891.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276652 | BC027810.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276652 | AK166833.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276652 | AK080772.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21769566 | - | - |
| Whole Mouse Genome G4122A | A_51_P276652 | BC052473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276652 | BC026891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276652 | BC027810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276652 | AK166833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P276652 | AK080772.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138108 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01392677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01392681 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769576 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769575 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769579 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21769582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21769582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01392677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21769584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01392681 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489000 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489002 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416928_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416929_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456964_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10488982 | - | - |
| Mouse Genome 430 2.0 Array | 1416929_at | - | - |
| Mouse Genome 430 2.0 Array | 1456964_at | - | - |
| Mouse Genome 430 2.0 Array | 1457934_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416929_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416928_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456964_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416928_at | - | - |
| Mouse Genome 430 2.0 Array | 1457934_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75710_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75710_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200042487.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042487.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042487.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042487.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042487.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042491.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042491.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042491.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042491.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042491.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042486.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042486.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042486.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042486.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042486.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042484.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042484.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042484.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042484.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042484.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042485.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042485.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042485.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042485.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042485.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042490.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042490.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042490.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042490.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042490.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042488.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042488.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042488.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042488.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042488.mm.1 | TC0200005511.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042489.mm.1 | NM_029397.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042489.mm.1 | NM_029397.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042489.mm.1 | NM_170598.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042489.mm.1 | NM_170598.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042489.mm.1 | TC0200005511.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416929_PM_at | - | - |
| Mouse Expression Set 430 | 1416928_PM_at | - | - |
| Mouse Expression Set 430 | 1416929_at | - | - |
| Mouse Expression Set 430 | 1416928_at | - | - |
| Mouse Expression Set 430 | 1456964_at | - | - |
| Mouse Expression Set 430 | 1456964_PM_at | - | - |
| Murine Genome U74v2 Set | 97199_PM_at | - | - |
| Murine Genome U74v2 Set | 97199_at | - | - |
| Murine Genome U74v2 Set | 169934_i_at | - | - |
| Murine Genome U74v2 Set | 169934_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416929_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416928_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416928_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416929_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416928_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416929_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416928_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416929_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456964_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456964_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660129 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4570239 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570239 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707710 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243646 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243646 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707710 | - | - |
| mouse-6 v1.1 (Illumina) | scl075710.1_90-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003069.1_215-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0075710.1_219-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040411 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660129 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570239 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003069.1_215-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0075710.1_219-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl075710.1_90-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570239 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660129 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221822 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2626300 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707710 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3104139 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221822 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3104139 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2626300 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707710 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0075710.1_219-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl075710.1_90-S | - | - |

Rbm12
Genomic Map