Assay Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029780.3 | NP_084056.1 | 2 - 3 | 635 | 68 | |
| GenBank mRNA | AB057655.1 | - | 1 - 2 | 180 | 68 | |
| AB057663.2 | - | 2 - 3 | 405 | 68 | ||
| AB077145.1 | - | 2 - 3 | 405 | 68 | ||
| AK036317.1 | - | 2 - 3 | 522 | 68 | ||
| AK141745.1 | - | 2 - 3 | 636 | 68 | ||
| BC015273.1 | - | 2 - 3 | 514 | 4222659 | 68 | |
| BC056439.1 | - | 2 - 3 | 554 | 68 | ||
| BC092040.1 | - | 2 - 3 | 500 | 2645313 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Raf1
Genomic Map