Assay Details
| Gene Symbol: Hook1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030014.2 | NP_084290.1 | 7 - 8 | 648 | 65 | |
| GenBank mRNA | AF487912.1 | - | 7 - 8 | 730 | 65 | |
| AK020924.1 | - | 7 - 8 | 672 | 65 | ||
| AK028714.1 | - | 7 - 8 | 667 | 65 | ||
| AK038941.1 | - | 7 - 8 | 659 | 65 | ||
| AK049897.1 | - | 7 - 8 | 649 | 65 | ||
| BC061688.1 | - | 7 - 8 | 648 | 5364426 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hook1
Genomic Map