Assay Details
| Gene Symbol: Hook1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030014.2 | NP_084290.1 | 13 - 14 | 1448 | 84 | |
| GenBank mRNA | AF487912.1 | - | 13 - 14 | 1530 | 84 | |
| AK028714.1 | - | 13 - 14 | 1467 | 84 | ||
| AK043870.1 | - | 13 - 14 | 1455 | 84 | ||
| AK049897.1 | - | 13 - 14 | 1449 | 84 | ||
| BC061688.1 | - | 13 - 14 | 1448 | 5364426 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hook1
Genomic Map