Assay Details
| Gene Symbol: Krt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033073.3 | NP_149064.1 | 7 - 8 | 1259 | 76 | |
| GenBank mRNA | AF509888.1 | - | 7 - 8 | 1236 | 76 | |
| AK002434.1 | - | 7 - 8 | 1237 | 76 | ||
| AK137217.1 | - | 1237 | 76 | |||
| AK146914.1 | - | 7 - 8 | 1237 | 76 | ||
| BC010337.1 | - | 6 - 7 | 3253 | 76 | ||
| BC030403.1 | - | 7 - 8 | 1270 | 5355662 | 76 | |
| BU755120.1 | - | 2 - 3 | 76 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Krt7
Genomic Map