Assay Details
| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033568.2 | NP_291046.1 | 6 - 7 | 658 | 103 | |
| GenBank mRNA | AK013058.1 | - | 6 - 7 | 660 | 103 | |
| AK020203.1 | - | 4 - 5 | 348 | 103 | ||
| BC003938.1 | - | 6 - 7 | 630 | 3486703 | 103 | |
| BC030490.1 | - | 1 - 2 | 623 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snf8
Genomic Map