Assay Details
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205343.1 | NP_001192272.1 | 4 - 5 | 320 | 121 | |
| NM_130887.3 | NP_570957.2 | 4 - 5 | 320 | 121 | ||
| GenBank mRNA | AF314171.1 | - | 4 - 5 | 293 | 121 | |
| AK053198.1 | - | 4 - 5 | 321 | 121 | ||
| AK086767.1 | - | 4 - 5 | 301 | 121 | ||
| AK138980.1 | - | 4 - 5 | 293 | 121 | ||
| BB664306.1 | - | 4 - 5 | 321 | 121 | ||
| BC132475.1 | - | 4 - 5 | 260 | 40130752 | 121 | |
| BC137854.1 | - | 4 - 5 | 260 | 8860873 | 121 | |
| BC144975.1 | - | 4 - 5 | 260 | 9053523 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Papln
Genomic Map