Assay Details
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205343.1 | NP_001192272.1 | 16 - 17 | 1869 | 80 | |
| NM_130887.3 | NP_570957.2 | 15 - 16 | 1803 | 80 | ||
| GenBank mRNA | AF314171.1 | - | 15 - 16 | 1776 | 80 | |
| AK053198.1 | - | 16 - 17 | 1870 | 81 | ||
| AK086767.1 | - | 15 - 16 | 1784 | 80 | ||
| AK138980.1 | - | 14 - 15 | 2668 | 80 | ||
| BC132475.1 | - | 15 - 16 | 1743 | 40130752 | 80 | |
| BC137854.1 | - | 15 - 16 | 1743 | 8860873 | 80 | |
| BC144975.1 | - | 16 - 17 | 1809 | 9053523 | 80 | |
More Information
Set Membership: |
|

Papln
Genomic Map