Assay Details
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205343.1 | NP_001192272.1 | 17 - 18 | 2124 | 103 | |
| NM_130887.3 | NP_570957.2 | 16 - 17 | 2058 | 103 | ||
| GenBank mRNA | AF314171.1 | - | 16 - 17 | 2031 | 103 | |
| AK053198.1 | - | 17 - 18 | 2126 | 103 | ||
| AK086767.1 | - | 16 - 17 | 2040 | 104 | ||
| AK138980.1 | - | 15 - 16 | 2924 | 103 | ||
| BC132475.1 | - | 16 - 17 | 1998 | 40130752 | 103 | |
| BC137854.1 | - | 16 - 17 | 1998 | 8860873 | 103 | |
| BC144975.1 | - | 17 - 18 | 2064 | 9053523 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Papln
Genomic Map