Assay Details
| Gene Symbol: Papln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205343.1 | NP_001192272.1 | 19 - 20 | 2390 | 100 | |
| NM_130887.3 | NP_570957.2 | 18 - 19 | 2324 | 100 | ||
| GenBank mRNA | AF314171.1 | - | 18 - 19 | 2297 | 100 | |
| AK053198.1 | - | 19 - 20 | 2392 | 100 | ||
| AK086767.1 | - | 18 - 19 | 2306 | 100 | ||
| BC132475.1 | - | 18 - 19 | 2264 | 40130752 | 100 | |
| BC137854.1 | - | 18 - 19 | 2264 | 8860873 | 100 | |
| BC144975.1 | - | 19 - 20 | 2330 | 9053523 | 100 | |
More Information
Set Membership: |
|

Papln
Genomic Map