Assay Details
| Gene Symbol: Hook2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167991.1 | NP_001161463.1 | 7 - 8 | 633 | 75 | |
| NM_133255.2 | NP_573518.2 | 7 - 8 | 633 | 75 | ||
| GenBank mRNA | AK030599.1 | - | 7 - 8 | 626 | 75 | |
| AK042048.1 | - | 7 - 8 | 621 | 75 | ||
| AK136069.1 | - | 7 - 8 | 627 | 75 | ||
| AK145768.1 | - | 7 - 8 | 601 | 75 | ||
| AK161612.1 | - | 7 - 8 | 586 | 75 | ||
| AK166954.1 | - | 7 - 8 | 613 | 75 | ||
| AK169073.1 | - | 7 - 8 | 601 | 75 | ||
| AK191641.1 | - | 3 - 4 | 173 | 75 | ||
| BC019486.1 | - | 7 - 8 | 609 | 4211990 | 75 | |
| BC029967.1 | - | 7 - 8 | 574 | 75 | ||
| BC055881.1 | - | 7 - 8 | 579 | 3590336 | 75 | |
| BC080744.1 | - | 7 - 8 | 623 | 30471369 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hook2
Genomic Map