Assay Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133701.2 | NP_598462.1 | 14 - 15 | 2055 | 70 | |
| GenBank mRNA | AK081998.1 | - | 14 - 15 | 1990 | 70 | |
| AK145461.1 | - | 14 - 15 | 2058 | 70 | ||
| AK166890.1 | - | 14 - 15 | 2007 | 70 | ||
| AK210126.1 | - | 3 - 4 | 353 | 70 | ||
| BC005801.1 | - | 4 - 5 | 571 | 70 | ||
| BC014869.1 | - | 14 - 15 | 1982 | 3602158 | 70 | |
| BC023691.1 | - | 14 - 15 | 1984 | 5344332 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prpf6
Genomic Map