Assay Details
| Gene Symbol: Prpf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133701.2 | NP_598462.1 | 17 - 18 | 2485 | 92 | |
| GenBank mRNA | AK004462.1 | - | 2 - 3 | 185 | 92 | |
| AK081998.1 | - | 17 - 18 | 2420 | 92 | ||
| AK145461.1 | - | 17 - 18 | 2488 | 92 | ||
| AK166890.1 | - | 17 - 18 | 2437 | 92 | ||
| BC005801.1 | - | 7 - 8 | 1001 | 92 | ||
| BC014869.1 | - | 17 - 18 | 2412 | 3602158 | 92 | |
| BC023691.1 | - | 17 - 18 | 2414 | 5344332 | 92 | |
| CJ228927.1 | - | 1 - 2 | 96 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf6
Genomic Map