Assay Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 19 - 20 | 2690 | 87 | |
GenBank mRNA | AK004462.1 | - | 4 - 5 | 389 | 86 | |
AK081998.1 | - | 19 - 20 | 2625 | 87 | ||
AK145461.1 | - | 19 - 20 | 2693 | 87 | ||
AK166890.1 | - | 19 - 20 | 2641 | 86 | ||
AK219156.1 | - | 2 - 3 | 147 | 87 | ||
BC005801.1 | - | 9 - 10 | 1206 | 87 | ||
BC014869.1 | - | 19 - 20 | 2617 | 3602158 | 87 | |
BC023691.1 | - | 19 - 20 | 2619 | 5344332 | 87 | |
CJ228927.1 | - | 3 - 4 | 301 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
