Assay Details
| Gene Symbol: Strn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039878.2 | NP_001034967.1 | 5 - 6 | 1205 | 63 | |
| NM_133789.3 | NP_598550.2 | 5 - 6 | 1205 | 63 | ||
| GenBank mRNA | AF414080.1 | - | 5 - 6 | 1205 | 63 | |
| AK136155.1 | - | 5 - 6 | 521 | 63 | ||
| AK154615.1 | - | 5 - 6 | 755 | 63 | ||
| AK161673.1 | - | 5 - 6 | 777 | 63 | ||
| AK197723.1 | - | 1 - 2 | 206 | 63 | ||
| BC004025.1 | - | 3 - 4 | 339 | 63 | ||
| BC080283.1 | - | 5 - 6 | 771 | 6401782 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strn4
Genomic Map