Assay Details
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139229.4 | NP_631975.3 | 4 - 5 | 1679 | 73 | |
| GenBank mRNA | AB041610.2 | - | 4 - 5 | 1602 | 73 | |
| AK083734.1 | - | 4 - 5 | 1621 | 73 | ||
| AK145515.1 | - | 4 - 5 | 1632 | 73 | ||
| AK145727.1 | - | 4 - 5 | 1621 | 73 | ||
| AK153617.1 | - | 4 - 5 | 1623 | 73 | ||
| AK156186.1 | - | 4 - 5 | 1622 | 73 | ||
| AK159569.1 | - | 4 - 5 | 1620 | 73 | ||
| BC023861.1 | - | 210 | 73 | |||
| BC094654.1 | - | 1615 | 6853777 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142832 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142834 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | BC094654.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK153617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK156186.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK159569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK083734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | BC094654.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK153617.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK156186.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK159569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P130824 | AK083734.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118188 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743133 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204206 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71173 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108721 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133282 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | BC094654.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK153617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK156186.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK159569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK083734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | BC094654.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK153617.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK156186.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK159569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P130824 | AK083734.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26142867 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108721 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01820657 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133282 | - | - |
| Whole Mouse Genome G4122A | A_51_P130824 | BC094654.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK153617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK156186.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK159569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK083734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | BC094654.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK153617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK156186.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK159569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P130824 | AK083734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P172145 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114009 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108721 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133282 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06820054 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06820047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07144348 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142833 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142836 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142837 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142834 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142879 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142838 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142877 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26142832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26142754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06971773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07277241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06781769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06971788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07170156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06971778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07083881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07269254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07181935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06781755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06781768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07220543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06781764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07181936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07220540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26142856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581510 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581512 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581511 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575143 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581513 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581517 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420265_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447860_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426821_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581507 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575142 | - | - |
| Mouse Genome 430 2.0 Array | 1420265_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447860_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1423617_at | - | - |
| Mouse Genome 430 2.0 Array | 1420265_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447860_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451052_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426821_at | - | - |
| Mouse Genome 430 2.0 Array | 1423617_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451052_at | - | - |
| Mouse Genome 430 2.0 Array | 1426821_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97484_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97484_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800022109.mm.1 | NM_026513.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022109.mm.1 | NM_026513.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022109.mm.1 | TC0800002869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022108.mm.1 | TC0800002869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022110.mm.1 | TC0800002869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022112.mm.1 | NM_139229.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022112.mm.1 | NM_139229.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022112.mm.1 | TC0800002869.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800022111.mm.1 | NM_139229.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022111.mm.1 | NM_139229.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800022111.mm.1 | TC0800002869.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451052_PM_at | - | - |
| Mouse Expression Set 430 | 1451052_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451052_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451052_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423617_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423617_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426821_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426821_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1420265_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447860_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1420265_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447860_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1230035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1230035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001230035 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226730 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2568092 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226730 | - | - |
| mouse-6 v1.1 (Illumina) | scl097484.1_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1230035 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl097484.1_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2595383 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2595383 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl097484.1_16-S | - | - |

Cog8
Genomic Map