Assay Details
| Gene Symbol: Sft2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145512.4 | NP_663487.1 | 6 - 7 | 537 | 84 | |
| GenBank mRNA | AK036578.1 | - | 6 - 7 | 425 | 84 | |
| AK075812.1 | - | 5 - 6 | 367 | 84 | ||
| AK156973.1 | - | 6 - 7 | 428 | 84 | ||
| AK170590.1 | - | 5 - 6 | 342 | 84 | ||
| BC017549.1 | - | 6 - 7 | 416 | 4924681 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sft2d2
Genomic Map