Assay Details
| Gene Symbol: Parp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145619.2 | NP_663594.2 | 4 - 5 | 664 | 66 | |
| GenBank mRNA | AF368233.1 | - | 4 - 5 | 664 | 66 | |
| AK147868.1 | - | 4 - 5 | 963 | 66 | ||
| AK170541.1 | - | 4 - 5 | 608 | 66 | ||
More Information
Set Membership: |
|

Parp3
Genomic Map