Assay Details
| Gene Symbol: Cd300lf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169153.1 | NP_001162624.1 | 1 - 2 | 154 | 65 | |
| NM_145634.3 | NP_663609.2 | 1 - 2 | 154 | 65 | ||
| GenBank mRNA | AB292061.1 | - | 1 - 2 | 43 | 65 | |
| AF251703.1 | - | 1 - 2 | 108 | 65 | ||
| AK089313.1 | - | 1 - 2 | 154 | 65 | ||
| AK089465.1 | - | 1 - 2 | 153 | 65 | ||
| AK154273.1 | - | 1 - 2 | 87 | 65 | ||
| AK155367.1 | - | 1 - 2 | 152 | 65 | ||
| AY214460.1 | - | 1 - 2 | 106 | 65 | ||
| AY457047.1 | - | 1 - 2 | 43 | 65 | ||
| BC057864.1 | - | 1 - 2 | 119 | 5318856 | 65 | |
| JX073136.1 | - | 1 - 2 | 129 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd300lf
Genomic Map