Assay Details
| Gene Symbol: Cd300lf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169153.1 | NP_001162624.1 | 2 - 3 | 501 | 83 | |
| GenBank mRNA | AF251703.1 | - | 2 - 3 | 455 | 83 | |
| AK089313.1 | - | 2 - 3 | 501 | 83 | ||
| AY457047.1 | - | 2 - 3 | 390 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd300lf
Genomic Map