Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145857.2 | NP_665856.2 | 3 - 4 | 640 | 107 | |
| GenBank mRNA | AF520774.1 | - | 3 - 4 | 685 | 107 | |
| AK089390.1 | - | 3 - 4 | 604 | 107 | ||
| AK089843.1 | - | 3 - 4 | 604 | 107 | ||
| AY160220.1 | - | 2 - 3 | 569 | 107 | ||
| AY160221.1 | - | 3 - 4 | 641 | 107 | ||
| DQ289012.1 | - | 2 - 3 | 162 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nod2
Genomic Map