Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145857.2 | NP_665856.2 | 5 - 6 | 2528 | 65 | |
| GenBank mRNA | AF520774.1 | - | 5 - 6 | 2573 | 65 | |
| AK089843.1 | - | 5 - 6 | 2492 | 65 | ||
| AY160220.1 | - | 4 - 5 | 2457 | 65 | ||
| AY160221.1 | - | 5 - 6 | 2529 | 65 | ||
| BC044774.2 | - | 5 - 6 | 2555 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nod2
Genomic Map