Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145857.2 | NP_665856.2 | 6 - 7 | 2615 | 75 | |
| GenBank mRNA | AF520774.1 | - | 6 - 7 | 2660 | 75 | |
| AK089843.1 | - | 6 - 7 | 2579 | 75 | ||
| AY160220.1 | - | 5 - 6 | 2544 | 75 | ||
| AY160221.1 | - | 6 - 7 | 2616 | 75 | ||
| BC044774.2 | - | 6 - 7 | 2642 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nod2
Genomic Map