Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145857.2 | NP_665856.2 | 8 - 9 | 2783 | 57 | |
| GenBank mRNA | AF520774.1 | - | 8 - 9 | 2828 | 57 | |
| AY160220.1 | - | 7 - 8 | 2712 | 57 | ||
More Information
Set Membership: |
|

Nod2
Genomic Map