Assay Details
| Gene Symbol: Nod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145857.2 | NP_665856.2 | 10 - 11 | 2958 | 78 | |
| GenBank mRNA | AF520774.1 | - | 10 - 11 | 3003 | 78 | |
| AK089843.1 | - | 10 - 11 | 2922 | 78 | ||
| AY160220.1 | - | 9 - 10 | 2887 | 78 | ||
| AY160221.1 | - | 10 - 11 | 2959 | 78 | ||
| BC044774.2 | - | 10 - 11 | 2985 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nod2
Genomic Map