Assay Details
| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153135.3 | NP_694775.1 | 6 - 7 | 1210 | 81 | |
| GenBank mRNA | AJ487854.1 | - | 6 - 7 | 1211 | 81 | |
| BC137615.1 | - | 6 - 7 | 1001 | 8860634 | 81 | |
| FJ824810.1 | - | 6 - 7 | 1143 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Unc5d
Genomic Map