Assay Details
| Gene Symbol: Stoml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153156.1 | NP_694796.1 | 1 - 2 | 36 | 71 | |
| GenBank mRNA | AB085692.1 | - | 1 - 2 | 36 | 71 | |
| BC058224.1 | - | 82 | 71 | |||
| BC080859.1 | - | 65 | 71 | |||
| BC138667.1 | - | 1 - 2 | 117 | 8861688 | 71 | |
| BC138668.1 | - | 1 - 2 | 117 | 8861689 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stoml3
Genomic Map