Assay Details
| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140324.1 | - | 2 - 3 | 447 | 77 | |
| AK157992.1 | - | 2 - 3 | 440 | 77 | ||
| AK165642.1 | - | 2 - 3 | 449 | 77 | ||
| BC030934.1 | - | 2 - 3 | 481 | 4920915 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox2
Genomic Map