Assay Details
| Gene Symbol: Zfp128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153802.4 | NP_722497.1 | 1 - 2 | 184 | 53 | |
| GenBank mRNA | AF480861.1 | - | 1 - 2 | 142 | 53 | |
| AK032046.1 | - | 1 - 2 | 165 | 53 | ||
| AK035906.1 | - | 1 - 2 | 185 | 53 | ||
| AK132244.1 | - | 1 - 2 | 186 | 53 | ||
| BC088998.1 | - | 1 - 2 | 155 | 6417536 | 53 | |
| BC094248.1 | - | 1 - 2 | 155 | 5718665 | 53 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK032046.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001558 | BC088998.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001558 | AK035906.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001558 | AK132244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001558 | AK032046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2001558 | NM_153802.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108873 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | AK053758.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | AK081042.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | AK157225.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | AK220337.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | BC072663.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | BC079565.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | BC079915.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 509621 | NM_001003912.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 883555 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 58910 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27505 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001558 | BC088998.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001558 | AK035906.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001558 | AK132244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001558 | AK032046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2001558 | NM_153802.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24973423 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01479773 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01479765 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06125562 | - | - |
| Whole Mouse Genome G4122A | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | BC088998.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | AK035906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | AK032046.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P104172 | NM_153802.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108579 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01479765 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06125562 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05974071 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05974077 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05979032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05979023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05979035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24973459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05908645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06162106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06001901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06001898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05974070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05896719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06161338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05896721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06192560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06192555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06192557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06086076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05974071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06086080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24973436 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550042 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550040 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458515_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439875_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550039 | - | - |
| Mouse Genome 430 2.0 Array | 1439875_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458515_at | - | - |
| Mouse Genome 430 2.0 Array | 1439875_at | - | - |
| Mouse Genome 430 2.0 Array | 1458515_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243833_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 243833_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700001122.mm.1 | TC0700000149.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001122.mm.1 | NM_153802.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001122.mm.1 | NM_153802.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001121.mm.1 | TC0700000149.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001121.mm.1 | NM_153802.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001121.mm.1 | NM_153802.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001123.mm.1 | TC0700000149.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700001123.mm.1 | NM_153802.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700001123.mm.1 | NM_153802.4 | Refseq |
| Mouse Expression Set 430 | 1458515_at | - | - |
| Mouse Expression Set 430 | 1439875_at | - | - |
| Mouse Expression Set 430 | 1458515_PM_at | - | - |
| Mouse Expression Set 430 | 1439875_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458515_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439875_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458515_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439875_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 545696 | - | - |
| ABI Mouse Genome Survey Microarray | 594953 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2060204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107050086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220117 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220117 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240884 | - | - |
| mouse-6 v1.1 (Illumina) | scl00243833.2_38-S | - | - |
| mouse-6 v1.1 (Illumina) | scl33080.4_549-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107050086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00243833.2_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060204 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107050086 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2828139 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2470238 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2828139 | - | - |

Zfp128
Genomic Map