Assay Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170671.2 | NP_733771.2 | 12 - 13 | 1668 | 91 | |
| GenBank mRNA | AK029929.1 | - | 11 - 12 | 1844 | 91 | |
| AK166780.1 | - | 5 - 6 | 816 | 91 | ||
| BC027327.1 | - | 8 - 9 | 1029 | 3991131 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mycbpap
Genomic Map