Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 7 - 8 | 543 | 85 | |
| GenBank mRNA | AK042692.1 | - | 7 - 8 | 544 | 85 | |
| AK044080.1 | - | 7 - 8 | 509 | 85 | ||
| AK047571.1 | - | 7 - 8 | 487 | 85 | ||
| AK137422.1 | - | 7 - 8 | 544 | 85 | ||
| BC115728.1 | - | 7 - 8 | 493 | 40039295 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne9
Genomic Map