Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 8 - 9 | 608 | 63 | |
| GenBank mRNA | AK042692.1 | - | 8 - 9 | 609 | 63 | |
| AK044080.1 | - | 8 - 9 | 574 | 63 | ||
| AK047571.1 | - | 8 - 9 | 552 | 63 | ||
| AK137422.1 | - | 8 - 9 | 609 | 63 | ||
| AK220550.1 | - | 1 - 2 | 1545 | 63 | ||
| BC115728.1 | - | 8 - 9 | 558 | 40039295 | 63 | |
| BE948663.1 | - | 5 - 6 | 63 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cpne9
Genomic Map