Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 11 - 12 | 860 | 75 | |
| GenBank mRNA | AK042692.1 | - | 11 - 12 | 861 | 75 | |
| AK044080.1 | - | 11 - 12 | 826 | 75 | ||
| AK047571.1 | - | 11 - 12 | 804 | 75 | ||
| AK137422.1 | - | 11 - 12 | 861 | 75 | ||
| AK220550.1 | - | 4 - 5 | 1797 | 75 | ||
| BC115728.1 | - | 11 - 12 | 810 | 40039295 | 75 | |
| BE948663.1 | - | 2 - 3 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cpne9
Genomic Map