Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 16 - 17 | 1281 | 65 | |
| GenBank mRNA | AK042692.1 | - | 16 - 17 | 1282 | 65 | |
| AK044080.1 | - | 16 - 17 | 1247 | 65 | ||
| AK047571.1 | - | 14 - 15 | 2296 | 65 | ||
| AK137422.1 | - | 16 - 17 | 1282 | 65 | ||
| AK220550.1 | - | 8 - 9 | 2645 | 65 | ||
| BC115728.1 | - | 16 - 17 | 1231 | 40039295 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne9
Genomic Map