Assay Details
| Gene Symbol: Hivep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007772.2 | NP_031798.2 | 3 - 4 | 273 | 140 | |
| GenBank mRNA | AK039808.1 | - | 3 - 4 | 273 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hivep1
Genomic Map