Assay Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077014.1 | - | 1340 | 104 | ||
| AK144176.1 | - | 8 - 9 | 1236 | 104 | ||
| BC005634.1 | - | 8 - 9 | 1210 | 104 | ||
| BC031757.1 | - | 7 - 8 | 1420 | 104 | ||
| BC046459.1 | - | 8 - 9 | 1349 | 5322519 | 104 | |
| BC057011.1 | - | 7 - 8 | 942 | 104 | ||
| BC071228.1 | - | 8 - 9 | 1292 | 4915902 | 104 | |
| X76858.1 | - | 7 - 8 | 1075 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

E4f1
Genomic Map