Assay Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF126967.1 | - | 9 - 10 | 1386 | 69 | |
| AK033285.1 | - | 9 - 10 | 1395 | 69 | ||
| AK144176.1 | - | 9 - 10 | 1343 | 69 | ||
| BC005634.1 | - | 9 - 10 | 1317 | 69 | ||
| BC031757.1 | - | 8 - 9 | 1527 | 69 | ||
| BC046459.1 | - | 9 - 10 | 1456 | 5322519 | 69 | |
| BC057011.1 | - | 8 - 9 | 1049 | 69 | ||
| X76858.1 | - | 8 - 9 | 1182 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

E4f1
Genomic Map