Assay Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF126967.1 | - | 12 - 13 | 1944 | 74 | |
| AK033285.1 | - | 12 - 13 | 1953 | 74 | ||
| AK077014.1 | - | 1980 | 74 | |||
| AK144176.1 | - | 12 - 13 | 1901 | 74 | ||
| BC005634.1 | - | 12 - 13 | 1875 | 74 | ||
| BC031757.1 | - | 11 - 12 | 2085 | 74 | ||
| BC046459.1 | - | 12 - 13 | 2014 | 5322519 | 74 | |
| BC057011.1 | - | 11 - 12 | 1607 | 74 | ||
| BC071228.1 | - | 12 - 13 | 1931 | 4915902 | 74 | |
| X76858.1 | - | 11 - 12 | 1740 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

E4f1
Genomic Map