Assay Details
| Gene Symbol: Ei24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199494.1 | NP_001186423.1 | 7 - 8 | 811 | 78 | |
| NM_007915.5 | NP_031941.1 | 7 - 8 | 695 | 78 | ||
| GenBank mRNA | AK145018.1 | - | 7 - 8 | 772 | 78 | |
| AK147776.1 | - | 7 - 8 | 724 | 78 | ||
| AK159927.1 | - | 7 - 8 | 753 | 78 | ||
| AK172233.1 | - | 7 - 8 | 775 | 78 | ||
| AK212828.1 | - | 2 - 3 | 256 | 78 | ||
| BU148129.1 | - | 7 - 8 | 764 | 78 | ||
| U41751.2 | - | 7 - 8 | 699 | 78 | ||
More Information
Set Membership: |
|

Ei24
Genomic Map