Assay Details
| Gene Symbol: Eps15l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122832.1 | NP_001116304.1 | 3 - 4 | 208 | 98 | |
| NM_001289859.1 | NP_001276788.1 | 3 - 4 | 208 | 98 | ||
| NM_007944.3 | NP_031970.2 | 3 - 4 | 208 | 98 | ||
| GenBank mRNA | AK036662.1 | - | 3 - 4 | 174 | 98 | |
| AK036728.1 | - | 3 - 4 | 186 | 98 | ||
| AK079132.1 | - | 3 - 4 | 176 | 98 | ||
| AK146781.1 | - | 3 - 4 | 186 | 98 | ||
| AK152602.1 | - | 3 - 4 | 186 | 98 | ||
| AK177985.1 | - | 2 - 3 | 133 | 98 | ||
| BC015259.1 | - | 3 - 4 | 189 | 4238768 | 98 | |
| BY270130.1 | - | 3 - 4 | 208 | 98 | ||
| U29156.1 | - | 3 - 4 | 195 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eps15l1
Genomic Map