Assay Details
| Gene Symbol: Evl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163394.1 | NP_001156866.1 | 6 - 7 | 1192 | 87 | |
| NM_001163395.1 | NP_001156867.1 | 6 - 7 | 944 | 87 | ||
| NM_001163396.2 | NP_001156868.1 | 7 - 8 | 966 | 87 | ||
| NM_007965.3 | NP_031991.3 | 6 - 7 | 1192 | 87 | ||
| GenBank mRNA | AF279662.1 | - | 5 - 6 | 703 | 87 | |
| AK134563.1 | - | 6 - 7 | 945 | 87 | ||
| AK151633.1 | - | 7 - 8 | 967 | 87 | ||
| BC059810.1 | - | 6 - 7 | 1192 | 6828523 | 87 | |
| BC096017.1 | - | 6 - 7 | 1085 | 87 | ||
| U72519.1 | - | 6 - 7 | 955 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Evl
Genomic Map